Merck
Severe reversible thrombocytopenia resulting from butoconazole cream.
DICP : the annals of pharmacotherapy (1990-02-01)
P A Maloley, E Nelson, H A Montgomery, J R Campbell
ZUSAMMENFASSUNG
A 54-year-old woman with a 19-year history of rheumatoid arthritis developed life-threatening thrombocytopenia one week after beginning butoconazole therapy for a vaginal yeast infection. This was complicated by upper gastrointestinal hemorrhage that probably resulted from ibuprofen and methotrexate therapy. Sepsis, myelophthisic anemias, and other potential etiologies were ruled out. Once stabilized, the patient was rechallenged with other medications without incident. These findings indicate that a potentially serious thrombocytopenia may result from the administration of butoconazole vaginal cream or in combination with methotrexate and/or ibuprofen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Butoconazolnitrat, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.