Merck
  • Multicolour optical coding from a series of luminescent lanthanide complexes with a unique antenna.
Multicolour optical coding from a series of luminescent lanthanide complexes with a unique antenna.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-01-29)
Nicolas Wartenberg, Olivier Raccurt, Elodie Bourgeat-Lami, Daniel Imbert, Marinella Mazzanti
ZUSAMMENFASSUNG
The bis-tetrazolate-pyridine ligand H(2)pytz sensitises efficiently the visible and/or near-IR luminescence emission of ten lanthanide cations (Pr, Nd, Sm, Eu, Tb, Dy, Ho, Er, Tm, Yb). The Ln(III) complexes present sizeable quantum yields in both domains with a single excitation source. The wide range of possible colour combinations in water, organic solvents and the solid state makes the complexes very attractive for labelling and encoding.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.