Merck
  • Inhibitory effects of in vivo oxidized high-density lipoproteins on platelet aggregation: evidence from patients with abetalipoproteinemia.
Inhibitory effects of in vivo oxidized high-density lipoproteins on platelet aggregation: evidence from patients with abetalipoproteinemia.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-03-20)
Catherine Calzada, Evelyne Véricel, Romain Colas, Nicolas Guillot, Graziella El Khoury, Jocelyne Drai, Agnès Sassolas, Noël Peretti, Gabriel Ponsin, Michel Lagarde, Philippe Moulin
ZUSAMMENFASSUNG
There is evidence that high-density lipoproteins (HDLs) may regulate platelet function, but disparate results exist regarding the effects of oxidized HDLs on platelets. The objective of our study was to determine the role of in vivo oxidized HDLs on platelet aggregation. Platelet aggregation and redox status were investigated in 5 patients with abetalipoproteinemia (ABLP) or homozygous hypobetalipoproteinemia, two rare metabolic diseases characterized by the absence of apolipoprotein B-containing lipoproteins, compared to 5 control subjects. Platelets isolated from plasma of patients with ABLP aggregated 4 to 10 times more than control platelets, depending on the agonist. By contrast, no differences in the extent of platelet aggregation were observed between ABLP platelet-rich plasma (PRP) and control PRP, suggesting the presence of a protective factor in ABLP plasma. ABLP HDLs inhibited agonist-induced platelet aggregation by binding to SR-BI, while control HDLs had no effect. On the other hand, lipoprotein-deficient plasma from patients with ABLP did not inhibit platelet aggregation. Severe oxidative stress was evidenced in patients with ABLP. Compared to control HDLs, ABLP HDLs showed a 40% decrease of α-tocopherol and an 11-fold increased malondialdehyde concentration. These results demonstrate that in vivo oxidized HDLs do not lose their antiaggregatory properties despite oxidation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
α-Tocopherylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-α-Tocopherolacetat, ≥96% (HPLC)
Sigma-Aldrich
Adenosin-5′-diphosphat, ≥95% (HPLC)
Sigma-Aldrich
Arachidonsäure, >95.0% (GC)
Supelco
DL-alpha-Tocopherolacetat, analytical standard
Sigma-Aldrich
Arachidonsäure, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
DL-α-Tocopherolacetat, tested according to Ph. Eur.
α-Tocopherol-acetat, European Pharmacopoeia (EP) Reference Standard
all-rac-α-Tocopherylacetat für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard