Merck
Fractionation and structural characterization of LiCl-DMSO soluble hemicelluloses from tomato.
Carbohydrate polymers (2013-04-03)
Carole Assor, Bernard Quemener, Jacqueline Vigouroux, Marc Lahaye
ZUSAMMENFASSUNG
To prepare and explore the structure of native hemicellulose from tomato, extraction of the natively acetylated polysaccharides was achieved from partially depectinated cell walls by DMSO doped with LiCl. DEAE anion exchange chromatography of the LiCl-DMSO extract allowed the removal of residual acidic pectin and the isolation of acetylated glucuronoxylan. The hemicellulose neutral fraction from the anion exchanger was fractionated by size exclusion chromatography into xyloglucan (XyG) and galactoglucomannan (GgM) either as single major constituents or as mixtures of both. Residual hemicellulose in the cell wall was extracted by 4.0 M and not 1.0 M KOH. The fine structure of all LiCl-DMSO fractions and alkali extracts was assessed by coupling β-glucanase, β-mannanase and β-xylanase enzymatic degradations to the analysis of the resulting fragments by HPAEC and MALDI-TOF mass spectrometry. This approach revealed substitutions in part of the GgM fractions by pentose residues, presumably arabinose and/or xylose occurring in highly substituted block domains. It also demonstrated a different glucanase hydrolysis profile from 4.0 M KOH compared to LiCl-DMSO soluble fractions. The present extraction and purification scheme allow the recovery of several populations of acetylated hemicellulose families which emphasize the structural diversity and complexity of these polysaccharides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Lithiumchlorid, ACS reagent, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Lithiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Lithiumchlorid -Lösung, 1 M in ethanol
Supelco
Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithiumchlorid, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithiumchlorid, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Xylanase aus Trichoderma viride, lyophilized powder, ≥100 units/mg protein
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®