Merck
Aegyptin inhibits collagen-induced coagulation activation in vitro and thromboembolism in vivo.
Biochemical and biophysical research communications (2013-06-04)
Daniella M Mizurini, Ivo M B Francischetti, Robson Q Monteiro
ZUSAMMENFASSUNG
Aegyptin is a mosquito salivary gland protein and potent inhibitor of platelet aggregation. Aegyptin binds to the von Willebrand factor-binding site on collagen and prevents its interaction with platelets. Because collagen also induces plasma clotting by activation of factor XII, we evaluated the effects of aegyptin on collagen-induced coagulation activation and how it interferes with thrombosis in three different in vivo models. Our results demonstrate that aegyptin abolishes collagen-induced clot formation and thrombin generation in platelet-free plasma. Aegyptin has no antithrombotic activity in the arteriovenous shunt model (collagen-independent) but it prevents laser-induced collagen-mediated thrombus formation in rats. Furthermore, aegyptin protects mice from collagen and epinephrine-induced thromboembolism. Therefore, aegyptin has a dual antithrombotic mechanism: inhibition of platelet-collagen interaction and collagen's pro-coagulant activity. This is the first description of a collagen-binding protein that also inhibits collagen-mediated coagulant activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Epinephrin
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Adrenalin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder