- [Microparasitocenosis of the intestines and the activity of intestinal enzymes in patients with trichocephaliasis during treatment].
[Microparasitocenosis of the intestines and the activity of intestinal enzymes in patients with trichocephaliasis during treatment].
Meditsinskaia parazitologiia i parazitarnye bolezni (1989-05-01)
N A Blagov, V N Firsov
PMID2779491
ZUSAMMENFASSUNG
Results of the studies of large intestine microflora, enterokinase and alkaline phosphatase activity in the feces of 298 children and adults suffering from trichocephaliasis are presented. Intestinal dysbacteriosis was observed in 51.7% cases, increased enterokinase activity, in 57.6% cases and increased alkaline phosphatase activity, in 55% cases. Enteric enzyme activity relation to the state of enteric microflora is demonstrated. Specific bephenium hydroxynaphthoate and mebendazole treatment was followed by increased dysbacteriosis and higher intestinal enzyme activity, especially in case of bephenium hydroxynaphthoate treatment. Normalization of the above-mentioned parameters was observed 90-120 days after the end of the treatment.