Merck
  • Profiles in altered metabolism. II--(omega -- 1)-hydroxyacid excretion in a case of episodic hypoglycemia.
Profiles in altered metabolism. II--(omega -- 1)-hydroxyacid excretion in a case of episodic hypoglycemia.
Biomedical mass spectrometry (1980-02-01)
O A Mamer, J A Montgomery, E Colle
ZUSAMMENFASSUNG
A patient with recurrent severe hypoglycemia resembling Reye's syndrome was found to have large accumulations of omega -- 1 hydroxy and keto acids in serum and urine that persisted following clinical recovery. A deficiency of mitochondrial medium chain acyl CoA dehydrogenase activity is proposed on the basis of evidence obtained using gas chromatographic mass spectrometric techniques. Analytical data is presented that will allow the recognition of ths variant presenting in other patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
7-Oxooctansäure, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.