Merck
  • Phorbol myristate acetate stimulates the formation of 5-oxo-6,8,11,14-eicosatetraenoic acid by human neutrophils by activating NADPH oxidase.
Phorbol myristate acetate stimulates the formation of 5-oxo-6,8,11,14-eicosatetraenoic acid by human neutrophils by activating NADPH oxidase.
The Journal of biological chemistry (1994-10-14)
W S Powell, F Gravelle, S Gravel
ZUSAMMENFASSUNG
We have shown previously that human neutrophil microsomes contain a highly specific dehydrogenase which, in the presence of NADP+, converts 5S-hydroxy-6,8,11,14-eicosatetraenoic acid (5S-HETE) to its 5-oxo metabolite, 5-oxo-ETE, a potent agonist of these cells. However, intact neutrophils convert 5S-HETE principally to its omega-oxidation product, 5,20-diHETE, and to only small amounts of 5-oxo-ETE. Phorbol myristate acetate (PMA) dramatically shifts the metabolism of 5S-HETE by intact cells so that 5-oxo-ETE is the major metabolite. The objective of this investigation was to determine the mechanism for the stimulatory effect of PMA on 5-oxo-ETE formation. The possibility that oxidants released in response to PMA nonenzymatically oxidized 5S-HETE was ruled out, since PMA did not appreciably stimulate the formation of 5-oxo-ETE from 5R-HETE. On the other hand, inhibition of NADPH oxidase either by diphenylene iodonium or by mild heating nearly completely prevented the stimulatory effect of PMA on the formation of 5-oxo-ETE. The possibility that this effect was mediated by superoxide seems unlikely, since it was still observed, although somewhat attenuated, in the presence of superoxide dismutase. Moreover, superoxide generated by another mechanism (xanthine/xanthine oxidase) did not appreciably affect the formation of 5-oxo-ETE by neutrophils. However, phenazine methosulfate, which can nonenzymatically convert NADPH to NADP+, mimicked the effect of PMA on 5-oxo-ETE formation by intact neutrophils. It is concluded that PMA acts by activating NADPH oxidase, resulting in conversion of NADPH to NADP+, which enhances the formation of 5-oxo-ETE and reduces the formation of 5,20-diHETE. Serum-treated zymosan has an effect on the metabolism of 5S-HETE similar to that of PMA in that it also stimulates the formation of 5-oxo-ETE and inhibits that of 5,20-diHETE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Prostaglandin B2, ≥98%, synthetic
Preise und Verfügbarkeit sind derzeit nicht verfügbar.