Merck
Active constituents against HIV-1 protease from Garcinia mangostana.
Planta medica (1996-08-01)
S X Chen, M Wan, B N Loh
ZUSAMMENFASSUNG
The ethanol extract of Garcinia mangostana L. (Guttiferae) showed potent inhibitory activity against HIV-1 protease. The activity-guided purification of the extract resulted in the isolation of two active, known compounds. The chemical structures of the isolated compounds were established by spectroscopic analyses as mangostin (IC50 = 5.12 +/- 0.41 microM) and gamma-mangostin (IC50 = 4.81 +/- 0.32 microM). The type of inhibition by both compounds is noncompetitive.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gamma-Mangostin, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.