Merck
  • PTH/PTHrP receptor is temporally regulated during osteoblast differentiation and is associated with collagen synthesis.
PTH/PTHrP receptor is temporally regulated during osteoblast differentiation and is associated with collagen synthesis.
Journal of cellular biochemistry (1996-06-15)
L K McCauley, A J Koh, C A Beecher, Y Cui, T J Rosol, R T Franceschi
ZUSAMMENFASSUNG
The temporal sequence of PTH/PTHrP receptor mRNA, binding, biologic activity, and its dependence on matrix synthesis was determined using MC3T3-E1 preosteoblast-like cells and primary rat calvarial cells in vitro. Osteoblastic cells were induced to differentiate and form mineralized nodules with the addition of ascorbic acid and beta-glycerophosphate, and samples were collected from 0-26 days of culture. DNA levels as determined by fluorometric analysis increased 12- and 17-fold during the collection period for both MC3T3-E1 and primary calvarial cells respectively. Steady state mRNA levels for the PTH/PTHrP receptor as determined by northern blot analysis, were initially low for both cell types, peaked at day 4 and 5 for MC3T3-E1 and primary calvarial cells respectively, and declined thereafter. Competition binding curves were performed during differentiation using 125I-PTHrP. The numbers of receptors per microgram DNA were greatest at days 3 and 5 for MC3T3-E1 and primary calvarial cells respectively. The biologic activity of the receptor was evaluated by stimulating the cells with 10 nM PTHrP and determining cAMP levels via a binding protein assay. The PTHrP-stimulated cAMP levels increased 5-fold to peak values at day 5 for MC3T3-E1! cells and 6-fold to peak values at day 4 for the primary calvarial cells. Ascorbic acid was required for maximal development of a PTH-dependent cAMP response since ascorbic acid-treated MC3T3-E1 cells had twice the PTH-stimulated cAMP levels as non-treated cells. When the collagen synthesis inhibitor 3,4-dehydroproline was administered to MC3T3-E1 cultures prior to differentiation, there was a subsequent diminution of the PTH/PTHrP receptor mRNA gene expression and numbers of receptors per cell; however, if administered after the initiation of matrix synthesis there was no reduction in PTH/PTHrP receptor mRNA. These findings indicate that the PTH/PTHrP receptor is associated temporally at the level of mRNA, protein, and biologic activity, with a differentiating, matrix-producing osteoblastic cell in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Dehydro-DL-Prolin, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.