- An endoglycosidase with alternative glycan specificity allows broadened glycoprotein remodelling.
An endoglycosidase with alternative glycan specificity allows broadened glycoprotein remodelling.
Journal of the American Chemical Society (2012-05-04)
Jonathan J Goodfellow, Kavitha Baruah, Keisuke Yamamoto, Camille Bonomelli, Benjamin Krishna, David J Harvey, Max Crispin, Christopher N Scanlan, Benjamin G Davis
PMID22551167
ZUSAMMENFASSUNG
Protein endoglycosidases are useful for biocatalytic alteration of glycans on protein surfaces, but the currently limited selectivity of endoglycosidases has prevented effective manipulation of certain N-linked glycans widely found in nature. Here we reveal that a bacterial endoglycosidase from Streptococcus pyogenes , EndoS, is complementary to other known endoglycosidases (EndoA, EndoH) used for current protein remodeling. It allows processing of complex-type N-linked glycans +/- core fucosylation but does not process oligomannose- or hybrid-type glycans. This biocatalytic activity now addresses previously refractory antibody glycoforms.