Merck
  • Improving the serum D-xylose test for the identification of patients with small intestinal malabsorption.
Improving the serum D-xylose test for the identification of patients with small intestinal malabsorption.
Journal of clinical gastroenterology (2001-06-22)
E D Ehrenpreis, M Salvino, R M Craig
ZUSAMMENFASSUNG
D-Xylose absorption testing is a simple, low-cost method of screening for small intestinal malabsorption. The optimum method to measure D-xylose absorption (serum vs. urine testing) is uncertain. We present a method of improving the accuracy of D-xylose testing. Fifty-one consecutive patients (40 with chronic diarrhea and 5 asymptomatic patients with renal insufficiency) and 6 volunteers with normal renal function were recruited. All received D-xylose, 10 g intravenously and 25 mg orally, on two separate occasions. Serum concentration was determined at baseline and at frequent times thereafter. Area under the curve was calculated to infinity, and D-xylose bioavailability (F) was calculated. A nonlinear model used to derive the relationship between 3-hour D-xylose concentrations and F showed that a value of less than 22.5 mg/dL correlated with an F of less than 60% (malabsorption of D-xylose). A 1-hour D-xylose of less than 20 mg/dL was considered abnormal. Using these indexes for normal 1-and 3-hour D-xylose levels, 90% of patients with D-xylose malabsorption were identified (sensitivity, 90%; specificity, 95%), which represents a marked improvement within the conventional 1-hour D-xylose of less than 20 mg/dL alone (sensitivity, 71%; specificity, 100%). The model was applied prospectively to 15 additional patients with chronic diarrhea. Of these, 12 patients with an F of less than 60% were identified, including 2 patients with normal 1-hour D-xylose levels. Thus, the addition of a 3-hour D-xylose serum level of less than 22.5 mg/dL to conventional 1-hour D-xylose determination greatly improves the D-xylose test for malabsorption screening.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Buttersäure, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-Mannitol, ACS reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aceton, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trimethylamin-N-oxid, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol, standard for GC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cadaverin, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.