Merck
  • Comparison of linuron degradation in the presence of pesticide mixtures in soil under laboratory conditions.
Comparison of linuron degradation in the presence of pesticide mixtures in soil under laboratory conditions.
Environmental monitoring and assessment (2013-03-26)
Maria Swarcewicz, Andrzej Gregorczyk, Justyna Sobczak
ZUSAMMENFASSUNG
It is widely recognised that complex interactions occur between chemicals in mixtures. In many agricultural situations, the use of tank mixes and complex spray programs is a common practice. Insecticides, fungicides and a herbicide being applied in potato protection were used in this research. Interactions between linuron and insecticides, such as thiamethoxam or clothianidin, and fungicides, such as mancozeb or chlorothalonil, were examined in soil. The degradation rate of linuron in soil during laboratory incubation in six treatments was studied. Mixtures of linuron with mancozeb in sandy loam and clay loam soils had a significant effect on the persistence of this herbicide. For example, for the same herbicide, t 1/2 values for linuron were from 37 days in sandy loam to 44 days in clay loam. These values changed (64-67 days) when thiamethoxam and mancozeb were in soil. When mancozeb was added only, the half-life values were from 59 to 62 days, respectively. Other mixtures with chlorothalonil, thiamethoxam and clothianidin did not have any effect. In order to compare linuron degradation rates in soils, a single first-order model and expanded statistical analysis were used.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Linuron, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.