Merck
  • Acute oral toxicity and liver oxidant/antioxidant stress of halogenated benzene, phenol, and diphenyl ether in mice: a comparative and mechanism exploration.
Acute oral toxicity and liver oxidant/antioxidant stress of halogenated benzene, phenol, and diphenyl ether in mice: a comparative and mechanism exploration.
Environmental science and pollution research international (2013-04-03)
Jiaqi Shi, Mingbao Feng, Xuesheng Zhang, Zhongbo Wei, Zunyao Wang
ZUSAMMENFASSUNG
The lethal doses (LD50s) of fluorinated, chlorinated, brominated, and iodinated benzene, phenol, and diphenyl ether in mice were ascertained respectively under the consistent condition. The acute toxicity of four benzenes orders in fluorobenzene (FB) < iodobenzene < chlorobenzene≈bromobenzene, that of four phenols orders in 4-iodophenol≈4-bromophenol < 4-chlorophenol (4-MCP) < 4-fluorophenol (4-MFP), and that of four diphenyl ethers orders in 4,4'-iododiphenyl ether < 4,4'-difluorodiphenyl ether < 4,4'-dichlorodiphenyl ether≈4,4'-dibromodiphenyl ether. General behavior adverse effects were observed, and poisoned mouse were dissected to observe visceral lesions. FB, 4-MCP, and 4-MFP produced toxic faster than other halogenated benzenes and phenols, as they had lower octanol-water partition coefficients. Pathological changes in liver and liver/kidney weight changes were also observed. Hepatic superoxide dismutase, catalase activities, and malondialdehyde level were tested after a 28-day exposure, which reflects a toxicity order basically consistent with that reflected by the LD50s. By theoretical calculation and building models, the toxicity of benzene, phenol, and diphenyl ether were influenced by different structural properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzol -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Diphenylether, ReagentPlus®, ≥99%
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol -Lösung, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Diphenylether, ≥99%, FG
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol -Lösung, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenylether, ReagentPlus®, 99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol -Lösung, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard