Merck
Flavoproteins are potential targets for the antibiotic roseoflavin in Escherichia coli.
Journal of bacteriology (2013-07-10)
Simone Langer, Masayuki Hashimoto, Birgit Hobl, Tilo Mathes, Matthias Mack
ZUSAMMENFASSUNG
The riboflavin analog roseoflavin is an antibiotic produced by Streptomyces davawensis. Riboflavin transporters are responsible for roseoflavin uptake by target cells. Roseoflavin is converted to the flavin mononucleotide (FMN) analog roseoflavin mononucleotide (RoFMN) by flavokinase and to the flavin adenine dinucleotide (FAD) analog roseoflavin adenine dinucleotide (RoFAD) by FAD synthetase. In order to study the effect of RoFMN and RoFAD in the cytoplasm of target cells, Escherichia coli was used as a model. E. coli is predicted to contain 38 different FMN- or FAD-dependent proteins (flavoproteins). These proteins were overproduced in recombinant E. coli strains grown in the presence of sublethal amounts of roseoflavin. The flavoproteins were purified and analyzed with regard to their cofactor contents. It was found that 37 out of 38 flavoproteins contained either RoFMN or RoFAD. These cofactors have different physicochemical properties than FMN and FAD and were reported to reduce or completely abolish flavoprotein function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Riboflavin (B2), analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
(−)-Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.