Merck
A modified cyclen azaxanthone ligand as a new fluorescent probe for Zn2+.
Dalton transactions (Cambridge, England : 2003) (2013-07-11)
Hela Nouri, Cyril Cadiou, Latévi Max Lawson-Daku, Andreas Hauser, Sylviane Chevreux, Isabelle Déchamps-Olivier, Fabien Lachaud, Riadh Ternane, Malika Trabelsi-Ayadi, Françoise Chuburu, Gilles Lemercier
ZUSAMMENFASSUNG
A new cyclen derivative L, bearing a methyl-chromeno-pyridinylidene hydrazone moiety, was synthesized and studied in MeOH, as potential fluorescent "OFF-on-ON" sensors for Zn(ii). Photophysical properties of this ligand being PET regulated, L was only weakly emissive in the absence of metal ions (OFF). L fluorescence was increased modestly upon addition of one equivalent of Zn(II), and further increased upon addition of a second equivalent. Therefore, Zn : L behaved as a highly sensitive ON sensor for zinc. This efficiency was correlated to Zn(II) coordination via the hydrazone moiety of the fluorophore, producing an efficient CHelation-Enhanced Fluorescence (CHEF) effect. A complementary theoretical study carried out with DFT calculations further elucidated the optical properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyclen, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.