Merck
  • In vivo dosimetry based on SPECT and MR imaging of 166Ho-microspheres for treatment of liver malignancies.
In vivo dosimetry based on SPECT and MR imaging of 166Ho-microspheres for treatment of liver malignancies.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2013-10-19)
Maarten L J Smits, Mattijs Elschot, Maurice A A J van den Bosch, Gerrit H van de Maat, Alfred D van het Schip, Bernard A Zonnenberg, Peter R Seevinck, Helena M Verkooijen, Chris J Bakker, Hugo W A M de Jong, Marnix G E H Lam, Johannes F W Nijsen
ZUSAMMENFASSUNG
(166)Ho-poly(l-lactic acid) microspheres allow for quantitative imaging with MR imaging or SPECT for microsphere biodistribution assessment after radioembolization. The purpose of this study was to evaluate SPECT- and MR imaging-based dosimetry in the first patients treated with (166)Ho radioembolization. Fifteen patients with unresectable, chemorefractory liver metastases of any origin were enrolled in this phase 1 study and were treated with (166)Ho radioembolization according to a dose escalation protocol (20-80 Gy). The contours of all liver segments and all discernible tumors were manually delineated on T2-weighted posttreatment MR images and registered to the posttreatment SPECT images (n = 9) or SPECT/CT images (n = 6) and MR imaging-based R2* maps (n = 14). Dosimetry was based on SPECT (n = 15) and MR imaging (n = 9) for all volumes of interest, tumor-to-nontumor (T/N) activity concentration ratios were calculated, and correlation and agreement of MR imaging- and SPECT-based measurements were evaluated. The median overall T/N ratio was 1.4 based on SPECT (range, 0.9-2.8) and 1.4 based on MR imaging (range, 1.1-3.1). In 6 of 15 patients (40%), all tumors had received an activity concentration equal to or higher than the normal liver (T/N ratio ≥ 1). Analysis of SPECT and MR imaging measurements for dose to liver segments yielded a high correlation (R(2) = 0.91) and a moderate agreement (mean bias, 3.7 Gy; 95% limits of agreement, -11.2 to 18.7). With the use of (166)Ho-microspheres, in vivo dosimetry is feasible on the basis of both SPECT and MR imaging, which enables personalized treatment by selective targeting of inadequately treated tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polymilchsäure, Mw ~60,000
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.