Merck
  • Sensitive detection of microcystin-LR by using a label-free electrochemical immunosensor based on Au nanoparticles/silicon template/methylene blue nanocomposite.
Sensitive detection of microcystin-LR by using a label-free electrochemical immunosensor based on Au nanoparticles/silicon template/methylene blue nanocomposite.
Journal of nanoscience and nanotechnology (2013-11-26)
Xuewen Fu, Yajuan Feng, Sipeng Niu, Chunling Zhao, Minghui Yang, Yunhui Yang
ZUSAMMENFASSUNG
A label-free electrochemical immunosensor, based on a gold nanoparticles (Au NPs)/silicon template/methylene blue (MB)/chitosan (CHIT) nanocomposite-modified electrode, was fabricated for the ultrasensitive detection of microcystin-LR (MC-LR). The nanocomposite film showed high binding affinity to the antibodies of MC-LR because gold nanoparticles have large surface area and good biocompatibility which can immobilize large amount of antibody through ionic interactions and other interactions between AuNPs and mercapto or primary amine groups of antibodies with high stability and bioactivity. MB was used as redox indicator due to its good electrochemical behavior in conductive substrate. This hybrid membrane was evaluated by cyclic voltammetry (CV), electrochemical impedance spectroscopy (EIS) and differential pulse voltammetry (DPV) to determine its electrochemical properties in immunosensor application. A decrease in DPV responses was observed with increasing concentrations of MC-LR in standard and real samples due to the formation of immuno-complexes between MC-LR and anti-MC-LR antibodies which hindered the electron charge transfer at the electrode-electrolyte interface. At optimal conditions, this immunosensor could detect MC-LR in a linear range from 0.5 ng/mL to 25 microg/mL with a low detection limit of 0.1 ng/mL at 3 sigma. Moreover, the prepared immunosensor was applied for the analysis of MC-LR in water samples with satisfactory results. The proposed method showed high selectivity, acceptable reproducibility, stability and reliability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Silizium, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Supelco
Microcystin-LR -Lösung, 10 μg/mL in methanol, analytical standard
Sigma-Aldrich
Methylenblau -Lösung, suitable for microscopy
Sigma-Aldrich
Methylenblau -Lösung, suitable for microbiology
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Silizium, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silizium, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm