Merck
  • [Characteristics of toxigenic properties of Myrothecium cinctum (Corda) sacc., 1886 and Myrothecium commune pidopl., 1969].
[Characteristics of toxigenic properties of Myrothecium cinctum (Corda) sacc., 1886 and Myrothecium commune pidopl., 1969].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2014-01-24)
A M Zaĭchenko, E S Tsyganenko, E V Andrienko, Ia I Savchuk
ZUSAMMENFASSUNG
The comparative study of toxigenic potential of Myrothecium cinctum (12 strains) and Myrothecium commune (7 strains) was conducted. The ability to produce the complex of macrocyclic trichothecenes (MCTC) presented in ethanol soluble fraction (ESF), as well as fungistatic activity against yeasts test-cultures (strains Saccharomyces vini "Feodosia" and Candida stellatoidea 62) and rabbit skin test were evaluated. It was established that ESF output varied within wide limits in all studied strains of both species but it was significantly lower in M. commune strains. Higher ESF yields were accompanied as a rule by higher fungistatic activity and skin reaction in rabbits. The separate strains showed high biological activity with minor ESF output that could be possible caused by domination of more toxic components in complex preparation. The study of MCTC component structure by high performance liquid chromatography method (HPLC) showed with a few exceptions the presence of verrucarins A and J, roridins A, E, and H as well as unidentified components whose column retention times were 2.1 and 4.9 minutes. The production of roridin H and unknown component with 2.1 min. retention time was typical of M. cinctum strains while verrucarin A and unidentified component with 4.9 min. retention time was typical of M. commune strains.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol, standard for GC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-200 (10 Ampullen/Kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-20 (10 Ampullen/Kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.