Merck
  • Synergistic photocatalytic degradation of pyridine using precious metal supported TiO2 with KBrO3.
Synergistic photocatalytic degradation of pyridine using precious metal supported TiO2 with KBrO3.
Journal of environmental sciences (China) (2014-02-21)
Fei Tian, Rongshu Zhu, Feng Ouyang
ZUSAMMENFASSUNG
A series of precious metals catalysts (M/TiO2, M = Ru, Rh, Pd, Ag, Ir, Pt or Au) were prepared by a light deposition method and the synergistic photocatalytic degradations of pyridine (20 mg/L) under UV irradiation (365 nm) using M/TiO2 with electron capture agent KBrO3 have been investigated. The results show that KBrO3 has a greatly synergistic role on M/TiO2 and the photocatalytic activity of M/TiO2 is closely related to its work function. Ag could greatly enhance the activity of TiO2 due to the binding characteristics of pyridine on Ag. Under the conditions of 0.5 wt.% Ag loading, Ag/TiO2 concentration of 0.1 g/L, KBrO3 concentration of 10 mmol/L and reaction liquid pH value at 9, the pyridine can be degraded by 64% within 3 hr, doubled than TiO2 photocatalytic system. The degradation kinetics of pyridine follows first-order kinetics and k = 5.53 x 10(-3) min-1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Titan(IV)-oxid, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, ReagentPlus®, ≥99%
Sigma-Aldrich
Titan(IV)-oxid, Anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titan, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Pyridin -hydrochlorid, 98%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Titan, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titan, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, ≥99.98% trace metals basis
Supelco
Pyridin, analytical standard
Sigma-Aldrich
Titan(IV)-oxid, Rutil, 99.995% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titan, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titan, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Pyridin -hydrobromid, 98%
Sigma-Aldrich
Titan, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titan, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titan, ≤10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)