- Molecular engineering of panchromatic isoindigo sensitizers for dye-sensitized solar cell applications.
Molecular engineering of panchromatic isoindigo sensitizers for dye-sensitized solar cell applications.
Chemical communications (Cambridge, England) (2014-03-19)
Shao-Gang Li, Ke-Jian Jiang, Jin-Hua Huang, Lian-Ming Yang, Yan-Lin Song
PMID24637700
ZUSAMMENFASSUNG
A panchromatic dye was synthesized with an isoindigo core as a linker to bridge with a bis(4-tert-butylphenyl)phenylamine donor and a cyanoacetic acid acceptor for dye-sensitized solar cells, showing a broad spectral response and a high conversion efficiency of 7.55% under AM 1.5 conditions.