Merck
  • [Ketoprophen and nefopam combination for postoperative analgesia with minimal use of narcotic analgesics in cardio-surgical patients].
[Ketoprophen and nefopam combination for postoperative analgesia with minimal use of narcotic analgesics in cardio-surgical patients].
Anesteziologiia i reanimatologiia (2014-03-15)
A A Eremenko, L S Sorokina, M V Pavlov
ZUSAMMENFASSUNG
4 combinations of analgesia were studied: 1) Nefopam and patient-controlled analgesia (PCA) with Trimeperidine; 2) Ketoprofen (100 microg each 12 hours intramuscular) and PCA with Trimeperidine; 3) Nefopam, Ketoprofen and PCA with Trimeperidine; 4) PCA with Trimeperidine as monotherapy in early postoperative period in cardio-surgical patients. 80 patients (age from 40 to 70) were divided into 4 groups, 20 patients in each group. Administration of Nefopam and Ketoprofen before extubation reduced the intensity of pain syndrome (in average on 90%) and promoted the early stirring up of patients. Combination of Nefopam and Ketoprofen provided the most expressed analgesic and opioids-saving effects. In this group average amount of Trimeperidine per 24 hours was 14.7 microg that was 4.9 times less than in group of PCA with Trimeperidine as monotherapy. Dynamics of maximal inspiratory capacity of the lungs in the first three groups was better than in group of PCA with Trimeperidine as monotherapy beginning from 6th hour of study. In common undesirable effects was connected with Trimeperidine administration and depended on its dose. The frequency of nausea, vomit, dizziness and weakness was authentically higher in the group of PCA with Trimeperidine as monotherapy than in other groups.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nefopam -hydrochlorid, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.