Merck
  • Stability and conformational change of methoxypolyethylene glycol modification for native and unfolded trypsin.
Stability and conformational change of methoxypolyethylene glycol modification for native and unfolded trypsin.
Food chemistry (2013-11-02)
Wei Liu, Jun-Ping Liu, Li-Qiang Zou, Zhao-Qin Zhang, Cheng-Mei Liu, Rui-Hong Liang, Ming-Yong Xie, Jie Wan
ZUSAMMENFASSUNG
The effect of succinimidyl carbonates activated methoxypolyethylene glycol (mPEG-SC) on the catalytic properties and conformation of native trypsin and dynamic high-pressure microfluidisation (DHPM) induced unfolded trypsin was studied. The thermal stability of unfolded trypsin was enhanced more significantly than that of native trypsin between 45 and 70°C. The autolysis analysis indicated that modified unfolded trypsin was markedly more resistant to autolysis compared to modified native trypsin between 40 and 180min. Upon mPEG-SC conjugation, the Km value of the enzyme decreased by about 2-fold, and the catalytic efficiency (Kcat/Km) increased by about 3-4-fold. Moreover, the increased thermal stability of unfolded trypsin might be due to the lower surface hydrophobicity and the higher hydrogen bond formation after mPEG-SC modification, which was reflected in the decrease of UV absorbance, the quenching and blue shift of fluorescence spectra, as well as the increase of β-sheet content.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypsin aus Schweinepankreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin aus Schweinepankreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin aus Schweinepankreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin aus Rinderpankreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Trypsin aus Rinderpankreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin aus Rinderpankreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin aus Schweinepankreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
TrypZean® Rind, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin aus Schweinepankreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin aus Schweinepankreas, ~1500 U/mg
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin aus meschlichem Pankreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin aus Rinderpankreas, ≥2,500 USP units/mg solid, meets USP testing specifications