Merck
The specificity of DNA recognition by the RAGE receptor.
The Journal of experimental medicine (2014-04-30)
Laure Yatime, Gregers Rom Andersen
ZUSAMMENFASSUNG
A recent paper by Sirois et al. in The Journal of Experimental Medicine reports that the receptor for advanced glycation end-products (RAGE) promotes uptake of DNA into endosomes and lowers the immune recognition threshold for the activation of Toll-like receptor 9. Two crystal structures suggested that the DNA phosphate-deoxyribose backbone is recognized by RAGE through well-defined interactions. However, the electron densities for the DNA molecules are weak enough that the presented modeling of DNA is questionable, and models only containing RAGE account for the observed diffraction data just as well as the RAGE-DNA complexes presented by the authors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Desoxyribonukleinsäure, einsträngig aus Lachshoden, For hybridization
Sigma-Aldrich
Desoxyribonukleinsäure, einsträngig aus Lachshoden, For hybridization
Sigma-Aldrich
Desoxyribonukleinsäure Natriumsalz aus Heringshoden, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Desoxyribonukleinsäure aus menschlicher Placenta, buffered aqueous solution, sexed, female