Merck
[Study progress on the mode of action of praziquantel against schistosomes].
Zhongguo ji sheng chong xue yu ji sheng chong bing za zhi = Chinese journal of parasitology & parasitic diseases (2008-04-30)
Shu-Hua Xiao
ZUSAMMENFASSUNG
Praziquantel is the only effective drug of choice against five human species of schistosomes. Main advantages of praziquantel include convenient oral administration, high safety and efficacy as well as short treatment course. To better understand the mode of action of praziquantel against schistosomes would be helpful for further development of new broad-spectrum anthelminthics. This paper summarizes the 30 years' research progress on the mode of action of the drug against schistosomes proceeded by domestic and abroad laboratories.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Praziquantel, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Praziquantel, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Praziquantel, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Praziquantel für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.