Merck
  • Meta-analysis: adefovir dipivoxil in combination with lamivudine in patients with lamivudine-resistant hepatitis B virus.
Meta-analysis: adefovir dipivoxil in combination with lamivudine in patients with lamivudine-resistant hepatitis B virus.
Virology journal (2009-10-13)
En-Qiang Chen, Li-Chun Wang, Jun Lei, Lu Xu, Hong Tang
ZUSAMMENFASSUNG
Currently, there are no conclusive results on the efficacy of adefovir dipivoxil (ADV) plus lamivudine (LAM) in LAM-resistant patients with chronic hepatitis B (CHB). The aim of study was to evaluate the efficacy of rescue therapy with ADV plus LAM compared to ADV monotherapy in LAM-resistant CHB patients. We searched PUBMED, EMBASE, Web of Science, CNKI (National Knowledge Infrastructure), VIP database, the Cochrane Central Register of Controlled Trials and the Cochrane Database of Systematic Reviews. Six eligible trials (442 patients in total) were included and evaluated for methodologic quality and heterogeneity. Greater virological response and lower emergence rate of ADV-associated mutants was observed in ADV plus LAM compared to ADV monotherapy (both P < 0.05). On the contrary, the rate of ALT normalization, HBeAg clearance and seroconversion were all similar between ADV plus LAM and ADV (all P > 0.05). Additionally, adding-on or switch-to ADV was both well tolerated. The combination of ADV with LAM was superior in inhibiting HBV replication and preventing drug resistance as compared to ADV alone for LAM-resistant CHB patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adefovir dipivoxil
Preise und Verfügbarkeit sind derzeit nicht verfügbar.