Merck
Dimethyl sulfoxide: an effective penetration enhancer for topical administration of NSAIDs.
The Physician and sportsmedicine (2011-10-28)
Karrie Marren
ZUSAMMENFASSUNG
Dimethyl sulfoxide (DMSO) is a molecule with a long history in pharmaceutics and is now well established as a penetration enhancer in topical pharmaceutical formulations. It is currently used for this purpose in diclofenac sodium topical solution (approved in the United States to treat signs and symptoms of osteoarthritis) and idoxuridine topical solution (approved in Europe for the treatment of herpes zoster). This article reviews the mechanism of action of DMSO as a pharmaceutical penetration enhancer, the characteristics of the molecule that facilitate transdermal drug delivery, and studies of efficacy and safety. The clinical use of pharmaceutical-grade DMSO as a penetration enhancer is supported by the robust data that have accumulated over the past 3 decades demonstrating the favorable safety and tolerability profile. Dimethyl sulfoxide is a safe and effective mechanism for facilitating the transdermal delivery of both hydrophilic and lipophilic medications to provide localized drug delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®