Merck
  • Six-month leuprorelin acetate depot formulations in advanced prostate cancer: a clinical evaluation.
Six-month leuprorelin acetate depot formulations in advanced prostate cancer: a clinical evaluation.
Clinical interventions in aging (2013-05-03)
Ulf W Tunn, Damian Gruca, Peter Bacher
ZUSAMMENFASSUNG
For nearly three decades, gonadotropin-releasing hormone (GnRH) agonists, particularly leuprorelin acetate (LA), have served as an important part of the treatment armamentarium for prostate cancer. The introduction of LA depot formulations provided a significant improvement in the acceptance of this therapy; however, their indicated treatment duration of 1 to 4 months was still not long enough to satisfy all medical needs. For this reason some manufacturers developed new injectable formulations that provide testosterone suppression for 6 months. This review article assesses key publications in order to compare these long-acting, commercially available, LA depot formulations and their clinical performance. The literature search identified 14 publications; by excluding reviews, duplications, and non-English articles, only three original papers describing clinical trial remained for review: two focused on microsphere-based LA formulations with either a 30 mg or 45 mg dose and one focused on a gel-based leuprorelin acetate with a 45 mg dose. All products were tested in individual clinical trials and have demonstrated their efficacy and safety.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Leuprorelin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.