Merck
Gadolinium contrast agent selection and optimal use for body MR imaging.
Radiologic clinics of North America (2014-06-04)
Flavius F Guglielmo, Donald G Mitchell, Shiva Gupta
ZUSAMMENFASSUNG
Proper selection of a gadolinium-based contrast agent (GBCA) for body magnetic resonance imaging (MRI) cases requires understanding the indication for the MRI exam, the key features of the different GBCAs, and the effect that the GBCA has on the selected imaging protocol. The different categories of GBCAs require timing optimization on postcontrast sequences and adjusting imaging parameters to obtain the highest T1 contrast. Gadoxetate disodium has many advantages when evaluating liver lesions, although there are caveats and limitations that need to be understood. Gadobenate dimeglumine, a high-relaxivity GBCA, can be used for indications when stronger T1 relaxivity is needed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gadolinium, chips
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.