Merck
Intestinal uptake and immunological effects of carrageenan--current concepts.
Food additives and contaminants (1989-10-01)
S Nicklin, K Miller
ZUSAMMENFASSUNG
Carrageenans are a group of high molecular weight sulphated polygalactans which find extensive use in the food industry as thickening, gelling and protein-suspending agents. Although there is no evidence to suggest that the persorption of small amounts of carrageenans across the intestinal barrier poses an acute toxic hazard, they are known to be biologically active in a number of physiological systems and extended oral administration in laboratory animals has been shown to modify both in vivo and in vitro immune competence. Whereas this effect could be attributed to carrageenan having a selective toxic effect on antigen-processing macrophages, additional studies suggest that macrophages can also influence immune responses by the timed release of immunoregulatory mediators. Evidence in support of this comes from in vitro studies which demonstrate that carrageenan-treated macrophages can, depending on conditions and time of administration, release either stimulatory or inhibitory factors. The former is known to be the immunostimulatory agent interleukin 1 (IL-1). The inhibitory factor, which is produced at an early stage following exposure to non-toxic doses of carrageenans, has yet to be formally identified but it is believed to be a prostaglandin because of its specific mode of action and short biological half-life. At present it is impossible to relate these studies to the human situation. Although it is established that carrageenans can cross the intestinal barrier of experimental animals, there is no evidence to suggest that the limited uptake that may occur in man in any way interferes with normal immune competence. Nevertheless, increased exposure may occur in the neonate during weaning, and adults and children following allergic reactions and episodes of gastrointestinal disease. Further studies under such conditions now seem warranted in order to elucidate the possible immunological consequences which may be associated with enhanced uptake of carrageenans in vulnerable groups.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carrageenan, suitable for gel preparation
Preise und Verfügbarkeit sind derzeit nicht verfügbar.