Merck
Repair of naphthalene-induced acute tracheal injury by basal cells depends on β-catenin.
The Journal of thoracic and cardiovascular surgery (2013-11-28)
Han-Shui Hsu, Chen-Chi Liu, Jiun-Han Lin, Tien-Wei Hsu, Kelly Su, Shih-Chieh Hung
ZUSAMMENFASSUNG
Little is known about the role of Wnt/β-catenin in postnatal airway homeostasis and basal cell function. This study aimed to investigate the role of Wnt signaling in the self-renewal of basal cells and the involvement of β-catenin in tracheal repair after naphthalene-induced injury. Mice were treated with naphthalene and injected with 4-hydroxytamoxifen. Injury and repair of the tracheal epithelium after naphthalene-mediated secretory cell depletion was assessed by a immunohistochemical study. The involvement of Wnt and β-catenin signaling in basal cell proliferation was investigated during in vitro expansion. Immunohistochemical analysis of tracheal epithelium in wild-type mice showed a reduction in the number of Clara cell secretory protein (CCSP+) and forkhead box transcription factor (Fox-J1+) cells on days 2 to 5 after naphthalene-induced injury; this cell population was regenerated by day 10. After flush labeling, bromodeoxyuridine-positive (BrdU+) cells and Ki67+ cells were observed in tracheal epithelium on days 2 to 5 but not on days 10 and 21. Confocal microscopy visualizing K5+ and BrdU+ cells showed that Wnt3a promotes proliferation of K5+ cells. Immunohistochemical analysis of K5+ and CCSP+ in tracheal epithelial cells from wild-type littermate and K5-Cre-mediated β-catenin knock-out mice showed that on day 3, the number of CCSP+ cells was decreased in all mice. On day 10, CCSP+ cells were present in wild-type littermate mice but absent in conditional knock-out mice. Basal cells serve as stem cells in the tracheal epithelium, regenerating and maintaining tracheal epithelial cells in a mouse model of tracheal injury. β-Catenin is required for proliferation and self-renewal of tracheal epithelial cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Naphthalin, 99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Naphthalin, analytical standard
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Supelco
Schmelzpunktstandard 79-81 °C, analytical standard
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Naphthalin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Naphthalin -Lösung, certified reference material, 5000 μg/mL in methanol
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Supelco
Naphthalin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®