Merck
Modulating endothelial barrier function by targeting vimentin phosphorylation.
Journal of cellular physiology (2014-03-22)
Tiegang Liu, Maher M Ghamloush, Ali Aldawood, Rod Warburton, Deniz Toksoz, Nicholas S Hill, Dale D Tang, Usamah S Kayyali
ZUSAMMENFASSUNG
Vimentin is a major intermediate filament protein in vascular endothelial cells which might be involved in their function as a barrier tissue. It is proposed to dynamically maintain integrity of the endothelium as a tightly regulated permeability barrier that is subjected to a variety of shear and contractile forces. The results described in this report demonstrate that vimentin plays that role through mechanisms that are dependent on its phosphorylation state. Withaferin A (WFA), a vimentin targeting drug is shown to disrupt endothelial barrier function through its effects on vimentin filament distribution and physical properties. These effects are related to WFA's ability to increase vimentin phosphorylation. Through overexpressing a non-phosphorylatable vimentin mutant we can block the effects of WFA on vimentin distribution and barrier permeability. The barrier augmentation effect appears to extend to endothelial cells that do not express detectable mutant vimentin which might suggest transmissible effects across cells. Blocking vimentin phosphorylation also protects the endothelial barrier against LPS endotoxin, implicating it as a target for drug development against pulmonary edema and acute respiratory distress syndrome (ARDS).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Withaferin A, ≥95% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Withaferin A, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Serin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.