Merck
  • Molecular cloning and expression analysis of a new bilin lyase: the cpcT gene encoding a bilin lyase responsible for attachment of phycocyanobilin to Cys-153 on the β-subunit of phycocyanin in Arthrospira platensis FACHB314.
Molecular cloning and expression analysis of a new bilin lyase: the cpcT gene encoding a bilin lyase responsible for attachment of phycocyanobilin to Cys-153 on the β-subunit of phycocyanin in Arthrospira platensis FACHB314.
Gene (2014-04-29)
Ran Zhang, Xiao-Ting Feng, Fei Wu, Yan Ding, Xiao-Nan Zang, Xue-Cheng Zhang, Ding-Yang Yuan, Bing-Ran Zhao
ZUSAMMENFASSUNG
To study the assembly of phycocyanin β subunit, the gene cpcT was first cloned from Arthrospira platensis FACHB314. To explore the function of cpcT, the DNA of phycocyanin β subunit and cpcT were transformed into Escherichia coli BL21 with the plasmid pET-hox1-pcyA, which contained the genes hemeoxygenase 1 (Hox1) and ferredoxin oxidoreductase (PcyA) needed to produce phycocyanobilin. The transformed strains showed specific phycocyanin fluorescence, and the fluorescence intensity was stronger than the strains with only phycocyanin β subunit, indicating that CpcT can promote the assembly of phycocyanin to generate fluorescence. To study the possible binding sites of apo-phycocyanin and phycocyanobilin, the Cys-82 and Cys-153 of the β subunit were individually mutated, giving two kinds of mutants. The results show that Cys-153 maybe the active site for β subunit binding to phycocyanobilins, which is catalyzed by CpcT in A. platensis FACHB314.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tributylzinnhydrid, contains 0.05% BHT as stabilizer, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
C-Phycocyanin from Spirulina sp., lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
[Arg8]-Vasopressin -acetat (Salz), ≥95% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 IU/mL in 0.9% NaCl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11, ≥97% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.