Merck
Citric acid assisted phytoremediation of cadmium by Brassica napus L.
Ecotoxicology and environmental safety (2014-05-21)
Sana Ehsan, Shafaqat Ali, Shamaila Noureen, Khalid Mahmood, Mujahid Farid, Wajid Ishaque, Muhammad Bilal Shakoor, Muhammad Rizwan
ZUSAMMENFASSUNG
Phytoextraction is an eco-friendly and cost-effective technique for removal of toxins, especially heavy metals and metalloids from contaminated soils by the roots of high biomass producing plant species with subsequent transport to aerial parts. Lower metal bioavailability often limits the phytoextraction. Organic chelators can help to improve this biological technique by increasing metal solubility. The aim of the present study was to investigate the possibility of improving the phytoextraction of Cd by the application of citric acid. For this purpose, plants were grown in hydroponics under controlled conditions. Results indicated that Cd supply significantly decreased the plant growth, biomass, pigments, photosynthetic characteristics and protein contents which were accompanied by a significant increase in Cd concentration, hydrogen peroxide (H₂O₂), electrolyte leakage, malondialdehyde (MDA) accumulation and decrease in antioxidant capacity. The effects were dose dependent with obvious effects at higher Cd concentration. Application of CA significantly enhanced Cd uptake and its accumulation in plant roots, stems and leaves. Citric acid alleviated Cd toxicity by increasing plant biomass and photosynthetic and growth parameters alone and in combination with Cd and by reducing oxidative stress as observed by reduction in MDA and H₂O₂ production and decreased electrolyte leakage induced by Cd stress. Application of CA also enhanced the antioxidant enzymes activity alone and under Cd stress. Thus, the data indicate that exogenous CA application can increase Cd uptake and minimize Cd stress in plants and may be beneficial in accelerating the phytoextraction of Cd through hyper-accumulating plants such as Brassica napus L.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Citronensäure, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
USP
Zitronensäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Zitronensäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Supelco
Zitronensäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Zitronensäure, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%