Merck
Vitamin B12 deficiency and bone health.
The New England journal of medicine (2014-09-04)
Thomas L Clemens
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taurin, ≥99%
Sigma-Aldrich
Vitamin B12, ≥98%
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Taurin, suitable for cell culture, meets USP testing specifications
Supelco
Cyanocobalamin (B12), analytical standard
Supelco
Vitamin B12, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Taurin, BioUltra, ≥99.5% (T)
Supelco
Taurin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Taurin, ≥98%, FG
SAFC
Taurin
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard