Merck
  • Pesticide multiresidue determination in rice paddy water by gas chromatography coupled with triple quadrupole mass spectrometry.
Pesticide multiresidue determination in rice paddy water by gas chromatography coupled with triple quadrupole mass spectrometry.
Journal of AOAC International (2014-08-26)
Mateus B Pereira, Janice F Facco, Gabriela M Zemolin, Manoel L Martins, Osmar D Prestes, Renato Zanella, Martha B Adaime
ZUSAMMENFASSUNG
Pollution of water sources by pesticides is a result of intensive use of these compounds, and the establishment of analytical methods for monitoring programs is very important. This paper presents a method for the determination of multiclass pesticide residues in rice paddy water based on SPE and GC/MS/MS. The validation results were satisfactory. All compounds showed adequate linearity (r2 >or= 0.99), and analysis by GC/MS/MS with a triple quadrupole mass spectrometer gave high selectivity and sensitivity. With the proposed SPE step an LOD of 0.06 microg/L was achieved. Recoveries from blank samples spiked at 0.2, 04, and 0.8 microg/L were between 70 and 120% with RSD <20% for most analyzed compounds, despite their different chemical nature, indicating good accuracy and precision. The proposed method is efficient for pesticide residue determination, including some metabolites, and gave good performance when applied to real samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
2,2,4-Trimethylpentan, ACS reagent, ≥99.0%
Supelco
Wasser
Sigma-Aldrich
2,2,4-Trimethylpentan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%