Merck
Integrin α3 mutations with kidney, lung, and skin disease.
The New England journal of medicine (2012-04-20)
Cristina Has, Giuseppina Spartà, Dimitra Kiritsi, Lisa Weibel, Alexander Moeller, Virginia Vega-Warner, Aoife Waters, Yinghong He, Yair Anikster, Philipp Esser, Beate K Straub, Ingrid Hausser, Detlef Bockenhauer, Benjamin Dekel, Friedhelm Hildebrandt, Leena Bruckner-Tuderman, Guido F Laube
ZUSAMMENFASSUNG
Integrin α(3) is a transmembrane integrin receptor subunit that mediates signals between the cells and their microenvironment. We identified three patients with homozygous mutations in the integrin α(3) gene that were associated with disrupted basement-membrane structures and compromised barrier functions in kidney, lung, and skin. The patients had a multiorgan disorder that included congenital nephrotic syndrome, interstitial lung disease, and epidermolysis bullosa. The renal and respiratory features predominated, and the lung involvement accounted for the lethal course of the disease. Although skin fragility was mild, it provided clues to the diagnosis.