Merck
  • Morphological Evolution of Single-Crystal Ag Nanospheres During the Galvanic Replacement Reaction with HAuCl4
Morphological Evolution of Single-Crystal Ag Nanospheres During the Galvanic Replacement Reaction with HAuCl<SUB>4</SUB>
The Journal of Physical Chemistry C (2008)
Kim MH, et al.