Merck
  • Multi-dimensional transition-metal coordination polymers of 4,4'-bipyridine-N,N'-dioxide: 1D chains and 2D sheets.
Multi-dimensional transition-metal coordination polymers of 4,4'-bipyridine-N,N'-dioxide: 1D chains and 2D sheets.
Inorganic chemistry (2008-08-30)
Junhua Jia, Alexander J Blake, Neil R Champness, Peter Hubberstey, Claire Wilson, Martin Schröder
ZUSAMMENFASSUNG
Reaction of 4,4'-bipyridine -N, N' -dioxide (L) with a variety of transition-metal salts in MeOH affords a range of coordination polymer products. For the complexes [FeCl 3(mu-L)] infinity, 1, and ([Cu(L) 2(OHMe) 2(mu-L)].2PF 6. n(solv)) infinity, 2, 1D chain structures are observed, whereas ([Mn(mu-L) 3].2ClO 4) infinity, 3, and ([Cu(mu-L) 3].2BF 4) infinity, 4, both show 2D sheet architectures incorporating an unusual 3 (6)- hxl topology. The more common 4 (4)- sql topology is observed in [Cd(ONO 2) 2(mu-L) 2] infinity, 5, ([Cu(OHMe) 2(mu-L) 2].2ZrF 5) infinity, 6, ([Cu(L) 2(mu-L) 2].2EF 6) infinity ( 7 E = P; 8 E = Sb), and ([Et 4N][Cu(OHMe) 0.5(mu-L) 2(mu-FSiF 4F) 0.5].2SbF 6. n(solv)) infinity, 9. In 6, the [ZrF 5] (-) anion, formed in situ from [ZrF 6] (2-), forms 1D anionic chains ([ZrF 5] (-)) infinity of vertex-linked octahedra, and these chains thread through a pair of inclined polycatenated ([Cu(OHMe) 2(mu-L) 2] (2+)) infinity 4 (4)- sql grids to give a rare example of a triply intertwined coordination polymer. 9 also shows a 3D matrix structure with 4 (4)- sql sheets of stoichiometry ([Cu(L) 2] (2+)) infinity coordinatively linked by bridging [SiF 6] (2-) anions to give a structure of 5-c 4 (4).6 (6)- sqp topology. The mononuclear [Cu(L) 6].2BF 4 ( 10) and [Cd(L) 6].2NO 3 ( 11) and binuclear complexes [(Cu(L)(OH 2)) 2(mu-L) 2)].2SiF 6. n(solv), 12, are also reported. The majority of the coordination polymers are free of solvent and are nonporous. Thermal treatment of materials that do contain solvent results in structural disintegration of the complex structures giving no permanent porosity.