Merck
  • Rebamipide attenuates autoimmune arthritis severity in SKG mice via regulation of B cell and antibody production.
Rebamipide attenuates autoimmune arthritis severity in SKG mice via regulation of B cell and antibody production.
Clinical and experimental immunology (2014-04-23)
J-K Byun, S-J Moon, J-Y Jhun, E-K Kim, J-S Park, J Youn, J-K Min, S-H Park, H-Y Kim, M-L Cho
ZUSAMMENFASSUNG
Oxidative stress is involved in the pathophysiology of rheumatoid arthritis (RA). We investigated the therapeutic potential of rebamipide, a gastroprotective agent with a property of reactive oxygen species scavenger, on the development of inflammatory polyarthritis and the pathophysiological mechanisms by which rebamipide might confer anti-arthritic effects in SKG mice, an animal model of RA. Intraperitoneal (i.p.) injection of rebamipide attenuated the severity of clinical and histological arthritis. Rebampide treatment reduced the number of T helper type 1 (Th1), Th2, Th17, inducible T cell co-stimulator (ICOS)(+) follicular helper T (Tfh) transitional type (T2) and mature B cells in the spleen, but increased the number of regulatory T (Treg ), CD19(+) CD1d(high) CD5(high) , CD19(+) CD25(high) forkhead box protein 3 (FoxP3)(+) regulatory B (Breg ) cells, memory B cells, and transitional type 1 (T1) B cells. In addition, flow cytometric analysis revealed significantly decreased populations of FAS(+) GL-7(+) germinal centre B cells and B220(-) CD138(+) plasma cells in the spleens of rebamipide-treated SKG mice compared to controls. Rebamipide decreased germinal centre B cells and reciprocally induced Breg cells in a dose-dependent manner in vitro. Rebamipide-induced Breg cells had more suppressive capacity in relation to T cell proliferation and also inhibited Th17 differentiation from murine CD4(+) T cells. Together, these data show that i.p. administration of rebamipide suppresses arthritis severity by inducing Breg and Treg cells and suppressing Tfh and Th17 cells in a murine model of RA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Alanin, BioUltra, ≥99.5% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Alanin, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
L-Alanin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Alanin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
L-Alanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Alanin-12C3, 99.9 atom % 12C
Preise und Verfügbarkeit sind derzeit nicht verfügbar.