Merck
  • The intracellular juxtamembrane domain of discoidin domain receptor 2 (DDR2) is essential for receptor activation and DDR2-mediated cancer progression.
The intracellular juxtamembrane domain of discoidin domain receptor 2 (DDR2) is essential for receptor activation and DDR2-mediated cancer progression.
International journal of cancer (2014-04-18)
Daehwan Kim, Panseon Ko, Eunae You, Sangmyung Rhee
ZUSAMMENFASSUNG
Discoidin domain receptors (DDRs) are unusual receptor tyrosine kinases (RTKs) that are activated by fibrillar collagens instead of soluble growth factors. DDRs play an important role in various cellular functions and disease processes, including malignant progression. Compared to other RTKs, DDRs have relatively long juxtamembrane domains, which are believed to contribute to receptor function. Despite this possibility, the function and mechanism of the juxtamembrane domain of DDRs have not yet been fully elucidated. In this study, we found that the cytoplasmic juxtamembrane 2 (JM2) region of DDR2 contributed to receptor dimerization, which is critical for receptor activation in response to collagen stimulation. A collagen-binding assay showed that JM2 was required for efficient binding of collagen to the discoidin (DS) domain. Immunohistochemical analysis of DDR2 expression using a tissue microarray demonstrated that DDR2 was overexpressed in several carcinoma tissues, including bladder, testis, lung, kidney, prostate and stomach. In H1299 cells, inhibition of DDR2 activity by overexpressing the juxtamembrane domain containing JM2 suppressed collagen-induced colony formation, cell proliferation and invasion via the inhibition of matrix metalloproteinase-2 and matrix metalloproteinase-9. Taken together, our results suggest that JM2-mediated dimerization is likely to be essential for DDR2 activation and cancer progression. Thus, inhibition of DDR2 function using a JM2-containing peptide might be a useful strategy for the treatment of DDR2-positive cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder