Merck
Comparing five simple vascular storage protocols.
The Journal of surgical research (2014-06-04)
Tristan P C van Doormaal, Jurren H Sluijs, Aryan Vink, Cornelis A F Tulleken, Albert van der Zwan
ZUSAMMENFASSUNG
We aim to find a storage protocol for vessels that preserves their dimensional, histologic, and mechanical characteristics to facilitate reproducible anastomosis experiments and microsurgical training with constant quality. We compared stored rabbit aortas, harvested in a slaughterhouse, using five different protocols with fresh controls. Aortas were preserved for 125 d in (1) NaCl 0.9% at -18°C, (2) Roswell Park Memorial Institute 1640 90% with 10% dimethyl sulfoxide (RPMI/DMSO) at -18°C, (3) RPMI/DMSO at -70°C, (4) glycerol 85% at 4°C, and (5) glycerol in stepwise increased concentrations until 85% at 4°C. After preservation, we measured vessel diameter, wall thickness, and Young's Modulus indicating stiffness. Neurosurgeons compared stored vessels with fresh vessels, blinded for preservation subgroup. We performed histologic assessment blinded for preservation subgroup. Fresh rabbit aortas showed a mean diameter of 2.65 ± 0.14 mm, a mean wall thickness of 126 ± 22 μm, and a Young's Modulus of 11.4 ± 2.4 N/mm(2). NaCl 0.9%-preserved aortas showed a significantly increased vessel diameter and decreased stiffness. RPMI/DMSO-preserved aortas showed no significant differences from fresh aortas in dimensions and mechanical characteristics. Glycerol-preserved tissue showed a significant increase in wall thickness, a related significant decrease in diameter, and increase in stiffness. Neurosurgeons regarded RPMI/DMSO tissue as most comparable with fresh tissue. Histologic assessment revealed no differences between the different protocols and fresh control group. Storage of rabbit aortas in RPMI/DMSO most adequately preserves their dimensional and mechanical properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis