Merck
  • Cardioprotective benefits of adenosine triphosphate-sensitive potassium channel opener diazoxide are lost with administration after the onset of stress in mouse and human myocytes.
Cardioprotective benefits of adenosine triphosphate-sensitive potassium channel opener diazoxide are lost with administration after the onset of stress in mouse and human myocytes.
Journal of the American College of Surgeons (2014-08-28)
M Burhan Janjua, Carol M Makepeace, Melissa M Anastacio, Richard B Schuessler, Colin G Nichols, Jennifer S Lawton
ZUSAMMENFASSUNG
Adenosine triphosphate-sensitive (KATP) potassium channel opener diazoxide (DZX) maintains myocyte volume and contractility during stress via an unknown mechanism when administered at the onset of stress. This study was performed to investigate the cardioprotective potential of DZX when added after the onset of the stresses of hyperkalemic cardioplegia, metabolic inhibition, and hypo-osmotic stress. Isolated mouse ventricular and human atrial myocytes were exposed to control Tyrode's solution (TYR) for 10 to 20 minutes, test solution for 30 minutes (hypothermic hyperkalemic cardioplegia [CPG], CPG + 100uM diazoxide [CPG+DZX], metabolic inhibition [MI], MI+DZX, mild hypo-osmotic stress [0.9T], or 0.9T + DZX), with DZX added after 10 or 20 minutes of stress, followed by 20 minutes of re-exposure to TYR (±DZX). Myocyte volume (human + mouse) and contractility (mouse) were compared. Mouse and human myocytes demonstrated significant swelling during exposure to CPG, MI, and hypo-osmotic stress that was not prevented by DZX when administered either at 10 or 20 minutes after the onset of stress. Contractility after the stress of CPG in mouse myocytes significantly declined when DZX was administered 20 minutes after the onset of stress (p < 0.05 vs TYR). Contractility after hypo-osmotic stress in mouse myocytes was not altered by the addition of DZX. To maintain myocyte volume homeostasis and contractility during stress (hyperkalemic cardioplegia, metabolic inhibition, and hypo-osmotic stress), KATP channel opener diazoxide requires administration at the onset of stress in this isolated myocyte model. These data have potential implications for any future clinical application of diazoxide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenolrot, powder, BioReagent, suitable for cell culture
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Phenolrot, ACS reagent
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.