Merck
Outcome of abdominal wall hernia repair with biologic mesh: Permacol™ versus Strattice™.
The American surgeon (2014-09-30)
Amy W Cheng, Maher A Abbas, Talar Tejirian
ZUSAMMENFASSUNG
The use of biologic mesh in abdominal wall operations has gained popularity despite a paucity of outcome data. Numerous biologic products are available with virtually no clinical comparison studies. A retrospective study was conducted to compare patients who underwent abdominal wall hernia repair with Permacol™ (crosslinked porcine dermis) and Strattice™ (noncrosslinked porcine dermis). Of 270 reviewed patients, 195 were implanted with Permacol™ and 75 with Strattice™. Ventral hernia repairs comprised the majority (85% for Permacol, 97% for Strattice™). Postoperative infection rate was lower in the Strattice™ group (5 vs 21%, P < 0.01). In the Permacol™ group only, the overall complication rates were significantly higher in patients with infected versus clean wounds (55 vs 35%, P < 0.05) and in obese patients (body mass index 40 kg/m(2) or greater [57 vs 34%], P < 0.01). Short-term complication and recurrence rates were higher when mesh was used as a fascial bridge: 51 versus 28 per cent for Permacol™, 58 versus 20 per cent for Strattice™. The hernia recurrence was similar in both groups. In this review of patients undergoing abdominal hernia repair with biologic mesh, Strattice™ mesh was associated with a lower short-term complication rate compared with Permacol™, but the hernia recurrence rate was similar.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder