Merck
  • MRIT, a novel death-effector domain-containing protein, interacts with caspases and BclXL and initiates cell death.
MRIT, a novel death-effector domain-containing protein, interacts with caspases and BclXL and initiates cell death.
Proceedings of the National Academy of Sciences of the United States of America (1997-10-23)
D K Han, P M Chaudhary, M E Wright, C Friedman, B J Trask, R T Riedel, D G Baskin, S M Schwartz, L Hood
ZUSAMMENFASSUNG
Activation of the cascade of proteolytic caspases has been identified as the final common pathway of apoptosis in diverse biological systems. We have isolated a gene, termed MRIT, that possesses overall sequence homology to FLICE (MACH), a large prodomain caspase that links the aggregated complex of the death domain receptors of the tumor necrosis factor receptor family to downstream caspases. However, unlike FLICE, the C-terminal domain of MRIT lacks the caspase catalytic consensus sequence QAC(R/Q)G. Nonetheless MRIT activates caspase-dependent death. Using yeast two-hybrid assays, we demonstrate that MRIT associates with caspases possessing large and small prodomains (FLICE, and CPP32/YAMA), as well as with the adaptor molecule FADD. In addition, MRIT simultaneously and independently interacts with BclXL and FLICE in mammalian cells. Thus, MRIT is a mammalian protein that interacts simultaneously with both caspases and a Bcl-2 family member.