Merck
Agitation-induced aggregation and subvisible particulate formation in model proteins.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-01-28)
Murali Jayaraman, Patrick M Buck, Arun Alphonse Ignatius, Kevin R King, Wei Wang
ZUSAMMENFASSUNG
The kinetics of agitation-induced subvisible particle formation was investigated for a few model proteins - human serum albumin (HSA), hen egg white lysozyme (HEWL), and a monoclonal antibody (IgG2). Experiments were carried out for the first time under relatively low protein concentration and low agitation speed to investigate the details of subvisible particle formation at the initial phase of aggregation (<2%) process. Upon agitation, both soluble higher molecular mass species (HMMS) and subvisible particles (SbVPs) formed at different rates, and via different mechanisms. Agitation enhanced exposure of hydrophobic sites in HSA but did not cause detectable structural changes in HEWL and IgG2. SbVPs from HSA partially dissociates in a neutral pH buffer (SEC mobile phase) but does not upon dilution in the same formulation buffer. Opposite results were obtained for SbVPs from IgG2 and HEWL. Neither the relative hydrophobic surface area nor the Tm of the model proteins seems to be an indicator of tendency for agitation-mediated SbVP formation. Taken together, our data suggests that agitation-induced SbVP formation can occur through different mechanisms and can vary, depending on the protein and solution conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
SYPRO Orange Protein-Gel-Farbstoff
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Albumin aus Humanserum, lyophilized powder, essentially fatty acid free
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®