Merck
  • Combination of immortalization and inducible death strategies to generate a human mesenchymal stromal cell line with controlled survival.
Combination of immortalization and inducible death strategies to generate a human mesenchymal stromal cell line with controlled survival.
Stem cell research (2014-02-25)
Paul Bourgine, Clementine Le Magnen, Sebastien Pigeot, Jeroen Geurts, Arnaud Scherberich, Ivan Martin
ZUSAMMENFASSUNG
The hTERT-immortalization of human bone marrow-derived Mesenchymal Stromal Cells (hMSCs) was proposed to address availability/standardization issues for experimental or clinical studies, but raised concerns due to possible uncontrolled growth or malignant cell transformation. Here we report a method to generate a hMSCs line with controlled survival, through the implementation of a pre-established suicide system (inducible caspase 9, iCasp9) in hTERT-transduced hMSCs. Primary hMSCs were successfully immortalized (>280 PD) and further transduced with the iCasp9 device. A clone was selected and shown to maintain typical properties of primary hMSCs, including phenotype, differentiation and immunomodulation capacities. The successive transductions did not induce tumorigenic transformation, as assessed by analysis of cell cycle regulators and in vivo luciferase-based cell tracking. Cells could be efficiently induced toward apoptosis (>95%) both in vitro and in vivo. By combining the opposite concepts of 'induced-life' and 'inducible-death', we generated a hMSCs line with defined properties and allowing for temporally controlled survival. The cell line represents a relevant tool for medical discovery in regenerative medicine and a potential means to address availability, standardization and safety requirements in cell & gene therapy. The concept of a hTERT-iCasp9 combination, here explored in the context of hMSCs, could be extended to other types of progenitor/stem cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hygromycin B aus Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Histochemisches Färbungskit für Seneszenzzellen, sufficient for 100 tests
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma