Merck
  • Metformin displays anti-myeloma activity and synergistic effect with dexamethasone in in vitro and in vivo xenograft models.
Metformin displays anti-myeloma activity and synergistic effect with dexamethasone in in vitro and in vivo xenograft models.
Cancer letters (2014-10-12)
Fu-Ming Zi, Jing-Song He, Yi Li, Cai Wu, Li Yang, Yang Yang, Li-Juan Wang, Dong-Hua He, Yi Zhao, Wen-Jun Wu, Gao-Feng Zheng, Xiao-Yan Han, He Huang, Qing Yi, Zhen Cai
ZUSAMMENFASSUNG
Epidemiologic studies and meta-analyses have suggested that patients with type 2 diabetes mellitus (T2DM) have a higher incidence of malignancies, including myeloma. Metformin is a widely prescribed antidiabetic drug. Recently, researchers have shown that metformin has direct anticancer activity against many tumor cell lines, mainly through activating AMP-activated protein kinase (AMPK) or reducing the blood insulin level. In the present study, we investigated whether metformin exerts an anti-myeloma effect in in vitro and in vivo xenograft models and explored the underlying mechanism. We found that metformin can inhibit proliferation of MM cells by inducing apoptosis and cell cycle arrest in the G0/G1 phase. Western blot showed that metformin activated caspase 3, caspase 9, PARP-1, Bak, and p21 and inactivated Mcl-1, HIAP-1, cyclin D1, CDK4, and CDK6. Metformin inhibited the expression of insulin growth factor-I receptor (IGF-IR), and phosphatidyl inositol 3-kinase (PI3K), protein kinase B (PKB/AKT) and the downstream mammalian target of rapamycin (mTOR). IGF-I blocked metformin-induced MM cell apoptosis and reactivation of the PI3K/AKT/mTOR signaling pathway. Metformin also demonstrated synergistic activity with dexamethasone but not bortezomib to eradicate MM cells in vitro and in vivo, especially in MM.1S cells. We conclude that metformin inhibits MM cell proliferation through the IGF-1R/PI3K/AKT/mTOR signaling pathway. Metformin and dexamethasone combination therapy may be an option for MM treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Dexamethason – wasserlöslich, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)