Merck
  • Ethanol-induced alcohol dehydrogenase E (AdhE) potentiates pneumolysin in Streptococcus pneumoniae.
Ethanol-induced alcohol dehydrogenase E (AdhE) potentiates pneumolysin in Streptococcus pneumoniae.
Infection and immunity (2014-10-15)
Truc Thanh Luong, Eun-Hye Kim, Jong Phil Bak, Cuong Thach Nguyen, Sangdun Choi, David E Briles, Suhkneung Pyo, Dong-Kwon Rhee
ZUSAMMENFASSUNG
Alcohol impairs the host immune system, rendering the host more vulnerable to infection. Therefore, alcoholics are at increased risk of acquiring serious bacterial infections caused by Streptococcus pneumoniae, including pneumonia. Nevertheless, how alcohol affects pneumococcal virulence remains unclear. Here, we showed that the S. pneumoniae type 2 D39 strain is ethanol tolerant and that alcohol upregulates alcohol dehydrogenase E (AdhE) and potentiates pneumolysin (Ply). Hemolytic activity, colonization, and virulence of S. pneumoniae, as well as host cell myeloperoxidase activity, proinflammatory cytokine secretion, and inflammation, were significantly attenuated in adhE mutant bacteria (ΔadhE strain) compared to D39 wild-type bacteria. Therefore, AdhE might act as a pneumococcal virulence factor. Moreover, in the presence of ethanol, S. pneumoniae AdhE produced acetaldehyde and NADH, which subsequently led Rex (redox-sensing transcriptional repressor) to dissociate from the adhE promoter. An increase in AdhE level under the ethanol condition conferred an increase in Ply and H2O2 levels. Consistently, S. pneumoniae D39 caused higher cytotoxicity to RAW 264.7 cells than the ΔadhE strain under the ethanol stress condition, and ethanol-fed mice (alcoholic mice) were more susceptible to infection with the D39 wild-type bacteria than with the ΔadhE strain. Taken together, these data indicate that AdhE increases Ply under the ethanol stress condition, thus potentiating pneumococcal virulence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol, standard for GC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-200 (10 Ampullen/Kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol-20 (10 Ampullen/Kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Ethanol -Lösung, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.