Merck
  • Dietary calcium deficiency and excess both impact bone development and mesenchymal stem cell lineage priming in neonatal piglets.
Dietary calcium deficiency and excess both impact bone development and mesenchymal stem cell lineage priming in neonatal piglets.
The Journal of nutrition (2014-10-17)
Yihang Li, Chad H Stahl
ZUSAMMENFASSUNG
Optimizing calcium nutrition to maximize bone accretion during growth to prevent fragility fractures later in life has spurred greater interest in calcium nutrition in neonates. The aim of this study was to determine the effect of dietary calcium, from deficiency through excess, on bone growth, and the in vivo and in vitro behavior of mesenchymal stem cells (MSCs) in neonatal pigs. Twenty-four male and female piglets (24 ± 6 h old) were fed either a calcium-deficient [Ca-D; 0.6% Ca on a dry matter (DM) basis], a calcium-adequate diet (Ca-A; 0.9% Ca on a DM basis), or a calcium-excessive diet (Ca-E; 1.3% Ca on a DM basis) for 14 d to assess the impact of dietary calcium on calcium homeostasis and on the behavior of MSCs. Growth rate was not affected by the Ca-E diet, although bone ash content was 16% higher (P < 0.05) and urinary calcium excretion was 5-fold higher, when normalized to creatinine, compared with the Ca-A group at trial completion. Serum parathyroid hormone (PTH) concentrations were elevated (P < 0.05) in Ca-D piglets in comparison with other groups at both 7 and 14 d. In vivo proliferation of MSCs was 30% higher (P < 0.05) in Ca-E piglets than the other groups. MSCs from both Ca-D- and Ca-E-fed piglets had greater adipogenic potential based on increased gene expression (P < 0.05) of peroxisome proliferator-activated receptor γ (Pparg) and adipocyte fatty acid-binding protein (Ap2) than MSCs from Ca-A piglets. Interestingly, only MSCs from Ca-E-fed piglets had greater (P < 0.05) gene expression of lipoprotein lipase (Lpl) during adipocytic differentiation than those from Ca-A piglets. To assess alterations in lineage allocation and priming, the most and least osteogenic (O+ and O-, respectively) and adipogenic (A+ and A-, respectively) colonies from each MSC isolation were selected on the basis of functional staining. The O+ colonies from Ca-D piglets expressed lower (P < 0.05) levels of osteocalcin (OC) mRNA than did those from other groups, whereas the O- colonies from Ca-E piglets expressed higher (P < 0.05) levels of mRNA of Pparg, Ap2, and Lpl than did those from other groups. Neonatal calcium deficiency appears to reduce the osteogenic priming of MSCs while enlarging a subpopulation of potentially adipogenic cells, and excess dietary calcium appears to allow greater multipotency of MSCs. These programming alterations of MSCs could have long-term consequences for bone health.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
D-Gluconsäure Natriumsalz, ≥99% (HPLC)
Sigma-Aldrich
Natriumgluconat, meets USP testing specifications
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
D-Gluconsäure Natriumsalz, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium D-Gluconat, 98.0-102.0%, meets USP testing specifications (Not intended for use in preparing injectable dosage forms.)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar